
2-(Allyloxy)benzaldehyde ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:28752-82-1
:
Formula:C₁₀H₁₀O₂
MDL Number:MFCD00014130
:162.19 g/mol
EINECS:249-198-2
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together