

4-(2-Chloroethyl)morpholine hydrochloride ≥97.0% (by titrimetric analysis)
Catalog # TCIAC1106-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (Nonaqueous Titration):min. 97.0 %
- Solubility in Water:very faint turbidity
Specification Test Results
About this item
:
:186.08 g/mol
EINECS:222-881-2
UN:2923
ADR:8,III
:GHS05
:GHS06
CAS Number:3647-69-6
Formula:C₆H₁₃Cl₂NO
MDL Number:MFCD00012797