Order Entry
Italy
ContactUsLinkComponent
2,5-Dibromo-3-dodecylthiophene ≥97.0% (by GC)
2,5-Dibromo-3-dodecylthiophene ≥97.0% (by GC)
Catalog # TCIAD4031-5G
Supplier:  TCI
CAS Number:  
undefined
2,5-Dibromo-3-dodecylthiophene ≥97.0% (by GC)
Catalog # TCIAD4031-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :410.25 g/mol
Storage Temperature:Refrigerator
CAS Number:148256-63-7
Formula:C₁₆H₂₆Br₂S
MDL Number:MFCD00274315