
4-tert-Butylcalix[5]arene ≥98.0% (by HPLC)
Catalog # TCIAB2221-100MG
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
About this item
:
:811.16 g/mol
:GHS07
CAS Number:81475-22-1
Formula:C₅₅H₇₀O₅
MDL Number:MFCD00210028