

(S)-(-)-1,1'-Bi-2-naphthol ≥98.0% (by HPLC, titration analysis)
: TCI
:18531-99-2
:
:C₂₀H₁₄O₂
:MFCD00004068
:286.33 g/mol
:2811
:6.1,III
:GHS06
:GHS08
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:207.0 to 212.0 deg-C
Frequently Bought Together