Order Entry
Italy
ContactUsLinkComponent
(R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone ≥98.0% (by HPLC, total nitrogen)
(R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
undefined
(R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Optical purity (LC):
    min. 99.0 ee%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    252.0 to 256.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together