Order Entry
Italy
ContactUsLinkComponent
(1-Naphthoxy)acetic acid ≥98.0% (by titrimetric analysis)
Irritant
(1-Naphthoxy)acetic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAN0046-10G
Supplier:  TCI
CAS Number:  
undefined
Irritant
(1-Naphthoxy)acetic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAN0046-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    194.0 to 198.0 deg-C

Specification Test Results

About this item

 :
 :202.21 g/mol
EINECS:221-023-4
 :GHS07
CAS Number:2976-75-2
Formula:C₁₂H₁₀O₃
MDL Number:MFCD00003927