Diethyl isopropylidenemalonate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6802-75-1
:
Formula:C₁₀H₁₆O₄
MDL Number:MFCD00009147
:200.23 g/mol
Boiling Pt:125°C
Flash Pt:80°C
EINECS:229-876-4
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together