
Di-tert-butyldichlorosilane ≥95.0% (by GC)
TCIAD2469-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
:
:213.22 g/mol
:82°C
:242-273-0
:2987
:8,II
:GHS05
:18395-90-9
:C₈H₁₈Cl₂Si
:MFCD00008796