Inserisci ordine
Italy
ContactUsLinkComponent
4,4-Dimethyl-2-oxazoline ≥97.0% (by GC, titration analysis)
Flammable
4,4-Dimethyl-2-oxazoline ≥97.0% (by GC, titration analysis)
Catalog # TCIAD2742-5ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
4,4-Dimethyl-2-oxazoline ≥97.0% (by GC, titration analysis)
Catalog # TCIAD2742-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Tipo di conf.:
    bottiglia in vetro
  • Conf:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %

Specification Test Results

About this item

 :
 :99.13 g/mol
Boiling Pt:100°C
Flash Pt:1°C
EINECS:250-045-7
UN:1993
ADR:3,II
 :GHS02
CAS Number:30093-99-3
Formula:C₅H₉NO
MDL Number:MFCD00014507