Order Entry
Italy
ContactUsLinkComponent
(±)-4-Methyl-1,3-dioxolane ≥98.0% (by GC)
Flammable
Irritant
(±)-4-Methyl-1,3-dioxolane ≥98.0% (by GC)
Catalog # TCIAM0916-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
(±)-4-Methyl-1,3-dioxolane ≥98.0% (by GC)
Catalog # TCIAM0916-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :88.11 g/mol
Boiling Pt:85°C
Flash Pt:-2°C
Flash Pt:–2°C
EINECS:214-008-9
UN:1993
ADR:3,II
 :GHS02
 :GHS07
CAS Number:1072-47-5
Formula:C₄H₈O₂
MDL Number:MFCD00014109