Order Entry
Italy
ContactUsLinkComponent
Sodium mercaptoacetate (sodium thioglycolate) ≥93.0% (by titrimetric analysis)
Irritant
Sodium mercaptoacetate (sodium thioglycolate) ≥93.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Sodium mercaptoacetate (sodium thioglycolate) ≥93.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Iodometric Titration):
    min. 93.0 %
  • Solubility in Water:
    almost transparency

Specification Test Results

Frequently Bought Together