

2-Methyl-1,3-dioxolane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:497-26-7
:
Formula:C₄H₈O₂
MDL Number:MFCD00003210
:88.11 g/mol
Boiling Pt:83°C
Flash Pt:–2°C
EINECS:207-841-4
UN:1993
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together