


4-Aminobutyraldehyde dimethyl acetal ≥98.0% (by GC, titration analysis)
Catalog # TCIAA1364-25ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
Boiling Pt:85°C
Flash Pt:59°C
UN:2924
ADR:3,III
:133.19 g/mol
EINECS:407-690-6
:GHS02
:GHS05
:GHS07
CAS Number:19060-15-2
Formula:C₆H₁₅NO₂
MDL Number:MFCD00189373