
2-Chloro-5-fluoropyrimidine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:62802-42-0
:
Formula:C₄H₂ClFN₂
MDL Number:MFCD03788197
:132.52 g/mol
Boiling Pt:94°C
Flash Pt:65°C
ADR:8,III
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together