
3-(4-Chlorobenzoyl)propionic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3984-34-7
:
Formula:C₁₀H₉ClO₃
MDL Number:MFCD00002794
:212.63 g/mol
EINECS:223-627-3
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:129.0 to 133.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together