1-Hydroxycyclohexyl phenyl ketone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: (1-Hydroxycyclohexyl)(phenyl)methanone
CAS Number:947-19-3
:
Formula:C₁₃H₁₆O₂
MDL Number:MFCD00059561
:204.27 g/mol
EINECS:213-426-9
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:47.0 to 50.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together