
3,3'-Dithiodipropionic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1119-62-6
:
Formula:C₆H₁₀O₄S₂
MDL Number:MFCD00002780
:210.28 g/mol
EINECS:214-284-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Melting point:155.0 to 159.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together