3-Dodecylthiophene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:104934-52-3
:
Formula:C₁₆H₂₈S
MDL Number:MFCD00130121
:252.46 g/mol
Boiling Pt:175°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together