

2-Chlorothiophene ≥98.0% (by GC)
Catalog # TCIAC0807-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:118.59 g/mol
Boiling Pt:128°C
Melting Pt:–1 °C
Density:1,285 g/cm³ (20 °C)
Flash Pt:23°C
Storage Temperature:Refrigerator
EINECS:202-505-3
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:96-43-5
Formula:C₄H₃ClS
MDL Number:MFCD00005421