
3-Bromo-2-chlorothiophene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:40032-73-3
:
Formula:C₄H₂BrClS
MDL Number:MFCD00043883
:197.48 g/mol
Boiling Pt:73°C
Density:1,82 g/cm³ (20 °C)
Flash Pt:102°C
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Colorless to Yellow to Orange clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together