

2,3-Dibromothiophene ≥95.0% (by GC)
: TCI
:3140-93-0
:
:C₄H₂Br₂S
:MFCD00005418
:241.93 g/mol
:219°C
:–17.5 °C
:2,2 g/cm³ (20 °C)
:51°C
:221-542-6
:1993
:3,III
:GHS02
:GHS07
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 95.0 %
Frequently Bought Together