

2,4,6-Collidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:108-75-8
:
Formula:C₈H₁₁N
MDL Number:MFCD00006338
Merck Index:14,09718
Boiling Pt:171°C
Melting Pt:–43 °C
Density:0,9191 g/cm³ (20 °C)
Flash Pt:58°C
UN:1992
ADR:3,III
:121.18 g/mol
EINECS:203-613-3
:GHS02
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light yellow to Red clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together