Order Entry
Italy
ContactUsLinkComponent
2,3-Lutidine ≥98.0% (by GC, titration analysis)
Flammable
Irritant
2,3-Lutidine ≥98.0% (by GC, titration analysis)
Catalog # TCIAL0063-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
2,3-Lutidine ≥98.0% (by GC, titration analysis)
Catalog # TCIAL0063-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :107.16 g/mol
Boiling Pt:161°C
Melting Pt:–15 °C
Density:0,945 g/cm³ (20 °C)
Flash Pt:50°C
EINECS:209-514-1
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:583-61-9
Formula:C₇H₉N
MDL Number:MFCD00009605