
2-(3-Phenylpropyl)pyridine ≥97.0% (by GC)
TCIAP1327-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
:
:197.28 g/mol
:171°C
:100°C
:218-300-7
:GHS07
:2110-18-1
:C₁₄H₁₅N
:MFCD00191237