Order Entry
Italy
ContactUsLinkComponent
2-Methoxypyridine ≥98.0% (by GC)
Flammable
Irritant
2-Methoxypyridine ≥98.0% (by GC)
Catalog # TCIAM0788-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
2-Methoxypyridine ≥98.0% (by GC)
Catalog # TCIAM0788-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :109.13 g/mol
Boiling Pt:142°C
Density:1,064 g/cm³ (0 °C)
Flash Pt:32°C
Storage Temperature:Ambient
EINECS:216-623-8
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:1628-89-3
Formula:C₆H₇NO
MDL Number:MFCD00006262