
3-Iodopyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1120-90-7
:
Formula:C₅H₄IN
MDL Number:MFCD00023553
:205 g/mol
Boiling Pt:92°C
Melting Pt:53.5 °C
EINECS:214-322-6
:GHS07
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:53.0 to 56.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together