
2,6-Dichloropyridine ≥97.0% (by GC)
: TCI
:2402-78-0
:
:C₅H₃Cl₂N
:MFCD00006244
:147.99 g/mol
:212°C
:83…86 °C
:Ambient
:219-282-3
:2811
:6.1,III
:GHS06
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:85.0 to 87.0 deg-C
- Solubility in Methanol:within almost transparency
Frequently Bought Together