
2,5-Dibromopyridine ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:624-28-2
:
Formula:C₅H₃Br₂N
MDL Number:MFCD00006221
:236.89 g/mol
Boiling Pt:235°C
Melting Pt:90…93 °C
Storage Temperature:Ambient
EINECS:210-839-6
:GHS07
Specification Test Results
- Appearance:White to Green to Brown powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:93.0 to 95.0 deg-C
Specification Test Results
Frequently Bought Together