
1,1'-Binaphthyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:604-53-5
:
Formula:C₂₀H₁₄
MDL Number:MFCD00041740
Boiling Pt:405 °C (1013 hPa)
Melting Pt:142…144 °C
Density:1,19 g/cm³ (20 °C)
:254.33 g/mol
EINECS:210-070-6
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Solubility in Toluene:very faint turbidity
Specification Test Results
Frequently Bought Together