Order Entry
Italy
ContactUsLinkComponent
1,3-Dimethylnaphthalene ≥95.0% (by GC)
Environment
1,3-Dimethylnaphthalene ≥95.0% (by GC)
Catalog # TCIAD0830-1G
Supplier:  TCI
CAS Number:  
undefined
Environment
1,3-Dimethylnaphthalene ≥95.0% (by GC)
Catalog # TCIAD0830-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
Boiling Pt:263°C
Melting Pt:–4 °C
Density:1,0146 g/cm³ (20 °C)
Flash Pt:109°C
Storage Temperature:Ambient
EINECS:209-384-6
UN:3082
ADR:9,III
 :156.23 g/mol
 :GHS09
CAS Number:575-41-7
Formula:C₁₂H₁₂
MDL Number:MFCD00004036