



1-Naphthol ≥98.0% (by GC) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:90-15-3
:
Formula:C₁₀H₈O
MDL Number:MFCD00003930
:144.17 g/mol
Merck Index:14,06383
Boiling Pt:147°C
Melting Pt:94 °C
Density:1,224 g/cm³ (20 °C)
Flash Pt:125 °C
EINECS:201-969-4
UN:2811
ADR:6.1,III
:GHS05
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Gray to Red powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:95.0 to 97.0 deg-C
- Peroxidase detection test:to pass test
Specification Test Results
Frequently Bought Together