2,6-Dimethoxynaphthalene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5486-55-5
:
Formula:C₁₂H₁₂O₂
MDL Number:MFCD00004063
:188.23 g/mol
EINECS:226-816-9
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:152.0 to 156.0 deg-C
- Solubility in hot Pyridine:almost transparency
Specification Test Results
Frequently Bought Together