
Octafluoronaphthalene ≥92.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:313-72-4
:
Formula:C₁₀F₈
MDL Number:MFCD00014307
:272.1 g/mol
Boiling Pt:209°C
Melting Pt:80…86 °C
Density:2,069 g/cm³ (-70 °C)
Storage Temperature:Ambient
EINECS:206-239-9
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 92.0 %
- Melting point:86.0 to 89.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
Frequently Bought Together