Order Entry
Italy
ContactUsLinkComponent
 
1-Naphthaldehyde ≥95.0% (by GC)
 :  TCI
 
undefined
1-Naphthaldehyde ≥95.0% (by GC)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Purity (GC):
    min. 95.0 %

 

Frequently Bought Together