
1-Bromonaphthalene ≥95.0% (by GC) stabilised
: TCI
:90-11-9
:
:C₁₀H₇Br
:MFCD00003868
:14,01425
:282°C
:0…6 °C
:1,48 g/cm³ (20 °C)
:66 °C
:207.07 g/mol
:201-965-2
:GHS07
:Ambient
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 %
Frequently Bought Together