

5,5-Diphenylhydantoin ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:57-41-0
:
Formula:C₁₅H₁₂N₂O₂
MDL Number:MFCD00005264
:252.27 g/mol
Merck Index:14,07322
EINECS:200-328-6
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Solubility in Acetone:almost transparency
Specification Test Results
Frequently Bought Together