

1,8-Diazabicyclo[5.4.0]undec-7-ene ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6674-22-2
:
Formula:C₉H₁₆N₂
MDL Number:MFCD00006930
:152.24 g/mol
Boiling Pt:83°C
Flash Pt:116°C
EINECS:229-713-7
UN:3267
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together