
1,1'-Dibromoferrocene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1293-65-8
:
Formula:C₁₀Br₂Fe
MDL Number:MFCD01075755
:335.76 g/mol
:GHS07
Storage Temperature:Refrigerata
Specification Test Results
- Appearance:Orange to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Chelometric Titration):min. 98.0 %
- Melting point:52.0 to 56.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together