5,5-Dimethylhydantoin ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:77-71-4
:
Formula:C₅H₈N₂O₂
MDL Number:MFCD00005266
:128.13 g/mol
EINECS:201-051-3
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:175.0 to 178.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together