



1,3-Dichloro-5,5-dimethylhydantoin ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:118-52-5
:
Formula:C₅H₆Cl₂N₂O₂
MDL Number:MFCD00003190
:197.02 g/mol
Merck Index:14,03065
EINECS:204-258-7
UN:3085
ADR:5.1,III
:GHS03
:GHS05
:GHS07
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Iodometric Titration):min. 97.0 %
- Melting point:132.0 to 136.0 deg-C
Specification Test Results
Frequently Bought Together