Order Entry
Italy
ContactUsLinkComponent
1,2-Epoxyhexadecane ≥80.0% (by GC)
Irritant
1,2-Epoxyhexadecane ≥80.0% (by GC)
Catalog # TCIAE0316-25ML
Supplier:  TCI
CAS Number:  
Irritant
1,2-Epoxyhexadecane ≥80.0% (by GC)
Catalog # TCIAE0316-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 80.0 %

Specification Test Results

About this item

 :
 :240.43 g/mol
Melting Pt:22°C
EINECS:230-786-2
 :GHS07
CAS Number:7320-37-8
Formula:C₁₆H₃₂O
MDL Number:MFCD00005151