


Benzyl chloroformate ≥96.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:501-53-1
:
Formula:C₈H₇ClO₂
MDL Number:MFCD00000640
:170.6 g/mol
Merck Index:14,01801
Boiling Pt:103°C
Melting Pt:0°C
Flash Pt:80°C
Storage Temperature:Refrigerator
EINECS:207-925-0
UN:1739
ADR:8,I
:GHS05
:GHS06
:GHS09
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 94.0 %
- Purity (Argentmetric Titration):min. 96.0 %
Specification Test Results
Related Information
Frequently Bought Together