


Sodium borohydride ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16940-66-2
:
Formula:NaBH₄
MDL Number:MFCD00003518
:37.83 g/mol
Merck Index:14,08592
Melting Pt:400 °C
Density:1,07 g/cm³ (20 °C)
Flash Pt:69 °C
Storage Temperature:Ambient
EINECS:241-004-4
UN:1426
ADR:4.3,I
:GHS02
:GHS05
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Sodium hypochlorite Method):min. 95.0 %
Specification Test Results
Related Information
Frequently Bought Together