Order Entry
Italy
Orders LinkContactUsLinkComponent
Methylcyclohexane ≥99.0% (by GC)

Methylcyclohexane ≥99.0% (by GC)

Flammable
Irritant
Health
Environment
Nome del fornitore:   TCI
Methylcyclohexane ≥99.0% (by GC)
Nome del fornitore:  TCI
Certificates
Methylcyclohexane ≥99.0% (by GC)

2 Options Available Below

Product Details & Documents
  • CAS Number:108-87-2
  • Formula:C₆H₁₁CH₃
  • MDL Number:MFCD00001497
  • Molecular Weight:98.19 g/mol
  • Merck Index:14,06047
  • Boiling Pt:100°C
  • Melting Pt:–127°C
  • Melting Pt:-127°C
  • Density:0,7693 g/cm³ (20 °C)
  • Flash Pt:–4°C
  • Flash Pt:-4°C
  • Storage Temperature:Ambient
  • EINECS:203-624-3
  • UN:2296
  • ADR:3,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Disponibilità
    Price
  • Glass bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 16,00 €
    16,00 €
    1 * 25 mL
       16,00 €

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 40,00 €
    40,00 €
    1 * 500 mL
       40,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.