2-Methyl-1,3-cyclopentanedione ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:765-69-5
:
Formula:C₆H₈O₂
MDL Number:MFCD00001406
:112.13 g/mol
Boiling Pt:213 °C (1013 hPa)
Density:1,33 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-153-2
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:210.0 to 217.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together