4-tert-Butylcyclohexanone ≥97.0% (by GC)
: TCI
:98-53-3
:
:C₁₀H₁₈O
:MFCD00001642
:154.25 g/mol
:116°C
:47…50 °C
:0,893 g/cm³ (50 °C)
:Ambient
:202-678-5
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:48.0 to 51.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together