
2-Amino-3-formylpyridine ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:7521-41-7
:
Formula:C₆H₆N₂O
MDL Number:MFCD01830382
:122.13 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:98.0 to 103.0 deg-C
Specification Test Results
Frequently Bought Together