
2-Bromo-3-formylpyridine ≥98.0% (by GC)
: TCI
:128071-75-0
:
:C₆H₄BrNO
:MFCD04966945
:186.01 g/mol
:100°C
:GHS07
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:71.0 to 75.0 deg-C
Frequently Bought Together