Order Entry
Italy
ContactUsLinkComponent
Thulium (III) trifluoromethanesulphonate ≥88.0% (by titrimetric analysis)
Corrosive
Thulium (III) trifluoromethanesulphonate ≥88.0% (by titrimetric analysis)
Catalog # TCIAT1920-5G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Thulium (III) trifluoromethanesulphonate ≥88.0% (by titrimetric analysis)
Catalog # TCIAT1920-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Chelometric Titration):
    min. 88.0 %

Specification Test Results

About this item

 :
ADR:8,III
 :616.14 g/mol
UN:1759
 :GHS05
CAS Number:141478-68-4
Formula:C₃F₉O₉S₃Tm
MDL Number:MFCD00209617